Learn More
Invitrogen™ MARK3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582890
Description
Immunogen sequence: QSPHHKVQRS VSSSQKQRRY SDHAGPAIPS VVAYPKRSQT STADSDLKED GISSRKSSGS AVGGKGIAPA SPMLGNASNP NKADIPERKK SSTVPSSNTA SGGMTRRNTY VCSERTTADR HSVIQNGKEN STIPDQRTPV ASTHSISSAA Highest antigen sequence indentity to the following orthologs: Mouse - 93%, Rat - 91%.
MARK3 is a member of a group of kinases known to regulate cell polarity via phosphorylation of microtubule-associated proteins (MAPs). The protein encoded by MARK3 is activated by phosphorylation and in turn is involved in the phosphorylation of tau proteins MAP2 and MAP4. Several transcript variants encoding different isoforms have been found for MARK3.
Specifications
| MARK3 | |
| Polyclonal | |
| Unconjugated | |
| MARK3 | |
| 1600015G02Rik; A430080F22Rik; cdc25C-associated protein kinase 1; cTAK1; C-TAK1; ELKL motif kinase 2; Emk2; EMK-2; EMK2/MARK3 serine-threonine protein kinase; ETK1; ETK-1; KP78; MAP/microtubule affinity regulating kinase 3; MAP/microtubule affinity-regulating kinase 3; MARK3; microtubule affinity regulating kinase 3; Mpk10; MPK-10; PAR1A; Par-1a; Protein kinase STK10; ser/Thr protein kinase PAR-1; Serine/threonine-protein kinase p78 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 170577, 17169, 4140 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P27448, Q03141, Q8VHF0 | |
| MARK3 | |
| Recombinant protein corresponding to Human MARK3. Recombinant protein control fragment (Product #RP-89020). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.