missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Mark3 Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | Mark3 |
|---|---|
| Dilution | Western Blot : 1-5 μg/mL, Immunoprecipitation |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30067006
|
Novus Biologicals™
NBP3-30800-250ug |
250 μg |
£386.00
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066947
|
Novus Biologicals™
NBP3-30800-50ug |
50 μg |
£162.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mark3 Polyclonal antibody specifically detects Mark3 in Human samples. It is validated for Immunoprecipitation,Western BlotSpecifications
| Mark3 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Sheep | |
| Protein Kinase | |
| Phosphate buffered saline | |
| 4140 | |
| IgG | |
| Ammonium sulfate precipitation |
| Western Blot : 1-5 μg/mL, Immunoprecipitation | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Cdc25C-associated protein kinase 1, cTAK1, C-TAK1, CTAK1ELKL motif kinase 2, EC 2.7.11, EC 2.7.11.1, EMK2, EMK-2, KP78, MAP/microtubule affinity-regulating kinase 3, PAR1A, Protein kinase STK10, Ser/Thr protein kinase PAR-1, Serine/threonine-protein kinase p78 | |
| Recombinant, full-length His-tagged human Mark3 produced in E. Coli. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title