missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15492-1-AP-20UL
This item is not returnable.
View return policy
Description
EMK (ELKL Motif Kinase) is a small family of ser/thr protein kinases involved in the control of cell polarity, microtubule stability and cancer. Several cDNA clones have been isolated that encoded two isoforms of the human ser/thr protein kinase EMK1. These isoforms were characterized by the presence of a 162-bp alternative exon that gave rise to two forms, one containing the exon and the other one lacking it. Both forms were found to be coexpressed in a number of selected cell lines and tissue samples. The human EMK1 was shown to be encoded by a single mRNA ubiquitously expressed.Specifications
| MARK2 | |
| Polyclonal | |
| Unconjugated | |
| MARK2 | |
| ELKL motif kinase 1, EMK 1, EMK1, MARK2, PAR 1, PAR1 homolog, Par1b | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13728, 2011, 60328 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O08679, Q05512, Q7KZI7 | |
| Mark2 | |
| MARK2 Fusion Protein Ag7884 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction