missing translation for 'onlineSavingsMsg'
Learn More

MARK2 Antibody, DyLight 405, Novus Biologicals™

Product Code. 18431394 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431394 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431394 Supplier Novus Biologicals Supplier No. NBP171890V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MARK2 Polyclonal antibody specifically detects MARK2 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MARK2
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.11, EC 2.7.11.1, ELKL motif kinase, ELKL motif kinase 1, EMK-1, EMK1PAR-1, MAP/microtubule affinity-regulating kinase 2MGC99619, PAR1 homolog, Par1b, Ser/Thr protein kinase PAR-1B, serine/threonine protein kinase EMK, serine/threonine-protein kinase MARK2
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 350 and 400 of human MAP/Microtubule Affinity-Regulating Kinase 2 using the numbering given in entry NP_001034558.2 (GeneID 2011).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2011
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.