missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Marimastat, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
Description
Marimastat (BB2516) is an angiogenesis and metastasis inhibitor, which limits the growth and production of blood vessels. As an antimetatstatic agent it prevents malignant cells from breaching the basement membranes.
Specifications
Specifications
| Chemical Name or Material | Marimastat |
| Physical Form | Solid |
| CAS | 154039-60-8 |
| Color | Off-White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C15H29N3O5 |
| For Use With (Application) | Cancer-Kinase/protease |
| Synonym | BB2516 TA2516 |
| Solubility Information | DMSO : 100 mg/mL (301.74 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction