missing translation for 'onlineSavingsMsg'
Learn More

Marimastat, MedChemExpress

Product Code. 16425008
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16425008

Brand: Medchem Express HY12169

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Marimastat (BB2516) is a broad spectrum and orally bioavailable inhibitor of MMPs, with potent activity against MMP-9 (IC50=3 nM), MMP-1 (IC50=5 nM), MMP-2 (IC50=6 nM), MMP-14 (IC50=9 nM) and MMP-7 (IC50=13 nM), used in the treatment of cancer.

Marimastat (BB2516) is an angiogenesis and metastasis inhibitor, which limits the growth and production of blood vessels. As an antimetatstatic agent it prevents malignant cells from breaching the basement membranes.

Specifications

Chemical Name or Material Marimastat
Physical Form Solid
CAS 154039-60-8
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C15H29N3O5
For Use With (Application) Cancer-Kinase/protease
Synonym BB2516 TA2516
Solubility Information DMSO : 100 mg/mL (301.74 mM; Need ultrasonic)
SMILES O=C(NO)[C@@H](O)[C@@H](CC(C)C)C(N[C@H](C(NC)=O)C(C)(C)C)=O
Molecular Weight (g/mol) 331.41
Formula Weight 331.41
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.