Learn More
Ansell Edmont™ Astroflex Immersion Gloves
Designed with high flexibility and excellent dexterity for chemical and liquid protection.
Brand:  Ansell Edmont™ ASTROFLEX_8
Product Code. 12990005
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Crinkle finish provides grip in wet and dry environments
  • Polyamide seamless liner
  • Wrinkle natural rubber latex coating
  • Gauntlet cuff