missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Maridebart Humanized Recombinant Human Monoclonal Antibody
Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA554936
This item is not returnable.
View return policy
Description
Compatible with Direct ELISA.
Maridebart works by inhibiting the enzyme dihydroorotate dehydrogenase (DHODH), which is essential for pyrimidine synthesis in rapidly dividing cells.
Specifications
| Maridebart Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| GIPR | |
| Recombinant Human GIPR/GIP-R. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA | |
| 0.92 mg/mL | |
| PBS with no preservative | |
| Human | |
| Protein A/G | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction