Learn More
Invitrogen™ Marginal Zone B Cells Monoclonal Antibody (HIS57), eBioscience™

Mouse Monoclonal Antibody
£226.00
Specifications
| Antigen | Marginal Zone B Cells |
|---|---|
| Clone | HIS57 |
| Concentration | 0.5 mg/mL |
| Content And Storage | 4°C |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunocytochemistry |
Description
Description: The monoclonal antibody HIS57 recognizes an unknown antigen expressed by rat marginal zone B cells. Marginal zone B cells are an anatomical B cell compartment located in the spleen at the area between red and white pulp. Marginal zone B cells serve as a first line of defense against blood-borne antigens in a T cell-independent manner. Marginal zone B cells can also be phenotyped as B cells that are IgM high, IgD low, CD21 high and CD23 low. Applications Reported: This HIS57 antibody has been reported for use in flow cytometric analysis, and immunohistochemical staining of frozen tissue sections. Applications Tested: This HIS57 antibody has been tested by immunohistochemistry of frozen rat tissue and can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
Marginal zone B cells are an anatomical B cell compartment located in the spleen at the area between red and white pulp. Marginal zone B cells serve as a first line of defense against blood-borne antigens in a T cell-independent manner. Marginal zone B cells can also be phenotyped as B cells that are IgM high, IgD low, CD21 high and CD23 low.Specifications
| Marginal Zone B Cells | |
| 0.5 mg/mL | |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Primary | |
| Affinity chromatography |
| HIS57 | |
| 4°C | |
| Monoclonal | |
| Liquid | |
| IgG1 κ | |
| Rat | |
| MZ-B; MZB1 | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.