missing translation for 'onlineSavingsMsg'
Learn More

SCAT™ Safety Funnel 'MARCO' with Lid, V2.0

Can be safely closed to prevent danger of ignition.

Brand:  SCAT™ 317639

231.78 GBP valid until 2024-12-31
Use promo code "23290" to get your promotional price.



Additional Details : Weight : 1.08900kg

 View more versions of this product

Product Code. 17863530

  • £287.00 / Each
Estimated Shipment: 14-06-2024
to see stock.
Add to basket

Description

Description

  • Modern lid funnel made of high-quality HDPE with very high resistance to acids, alkalis and many other chemicals
  • Integrated gripping edge on funnel lid increases grip without having to touch possibly contoured lid
  • Innovative funnel lid with integrated sealing ring closes funnel edge to prevent escape of explosive or health-endangering gases
  • Sturdy hinge guarantees long service life of the closing mechanism
  • Rotating screw cap makes it easier to screw lid funnel on and off the canister
  • Removable sieve for easy cleaning
  • Conductive version of MARCO V2.0 is black, differing in color from the non-conductive version
Specifications

Specifications

Safety Funnel with Hinged Lid
HDPE, Conductive
Black
S70/71
140 mm
Not Autoclavable
Funnel with Lid, Removable Sieve, Earthing Cable, Level Control with 150 mm Floater
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers