missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARCO Rabbit anti-Human, Clone: PLK1, Novus Biologicals™
Rabbit Monoclonal Antibody
£209.00 - £463.00
Specifications
| Antigen | MARCO |
|---|---|
| Clone | PLK1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18323862
|
Novus Biologicals
NBP3-12048-0.025MG |
0.025 mg |
£209.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18048135
|
Novus Biologicals
NBP3-12048-0.2MG |
0.2 mg |
£463.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MARCO Monoclonal antibody specifically detects MARCO in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen)Specifications
| MARCO | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| RUO | |
| PBS | |
| 8685 | |
| IgG κ | |
| Protein A purified |
| PLK1 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen | |
| Monoclonal | |
| Rabbit | |
| Human | |
| macrophage receptor with collagenous structureSCARA2Scavenger receptor class A member 2, member 2 | |
| The antibody was raised by immunising BALB/c mice with human alveolar macrophages (AMs) and subsequent creation of hybridomas. | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title