Learn More
Emma Safety Footwear Marco Frontier S3 Shoe
Product Code. 17562436
Change view
Click to view available options
Closure Type:
Laces
Unit Size:
1 pair
Product Code. Closure Type Unit Size
17562436 Laces 1 pair
1 options
Product Code. 17562436 Supplier Emma Safety Footwear Supplier No. MM11209045
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Break new ground in the Marco Frontier, the executive safety shoe that literally has no boundaries.
  • Industries
    • Construction & technology
    • Manufacturing
    • Government & services
    • Oil & gas
    Safety shoe
    • S3-Safetyshoe with sole plate for wet work
Specifications
Closure Type Laces
Color Brown
Material Leather
Size 45
Toe Material Steel
Fit Standard Width
Ankle Style Low-cut
Product Type Safety Shoe