Learn More
Emma Safety Footwear Marco (Frontier 112) Safety Shoes
Product Code. 15978181
Change view
Click to view available options
Size:
39
40
41
42
43
44
46
47
48
Unit Size:
1 pair
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15978181 39 1 pair
15988181 40 1 pair
15612469 41 1 pair
15775233 42 1 pair
15865771 43 1 pair
15785233 44 1 pair
15998181 46 1 pair
15908191 47 1 pair
15918191 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 15978181 Supplier Emma Safety Footwear Supplier No. MM11209039
Only null left
Add to basket
Add to basket
Made with the same care and  attention to detail associated with  high end luxury shoes. Brazilian 2 tone full grain leather combined with nubuck adds to the stylish look.
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • Thermoplastic Polyurethane Outsole
  • ESD certified
  • Chrome-free lining
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20345:2011, EN 12568:2010
Closure Type Laces
Color Brown
Design Marco (Frontier 112)
Liner Material Hydro-Tec™ Sanitized Silver & Wellness leather
Material Leather
Size 39
Toe Material Steel