missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Marcksl1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10002-2-AP-20UL
This item is not returnable.
View return policy
Description
Controls cell movement by regulating actin cytoskeleton homeostasis and filopodium and lamellipodium formation. When unphosphorylated, induces cell migration. When phosphorylated by MAPK8, induces actin bundles formation and stabilization, thereby reducing actin plasticity, hence restricting cell movement, including neuronal migration. May also affect cancer cell migration. May be involved in coupling the protein kinase C and calmodulin signal transduction systems.Specifications
| Marcksl1 | |
| Polyclonal | |
| Unconjugated | |
| MARCKSL1 | |
| AL022768, AW215397, AW536807, D4Bc1, F52, MacMARCKS, Macs 2, Macs 3, Macs2, Macs3, MARCKS like 1, Marcksl1, Mlp, Mrp | |
| Rabbit | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P28667, P49006 | |
| Marcksl1 | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17357, 65108 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction