missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MARCKSL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103304
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MRP.
MARCKS (Myristoylated alanine-rich protein kinase C substrate), also designated 80K or 80K-L, is a 332 amino acid protein that localizes to the plasma membrane and functions as a major cellular substrate for protein kinase C (PKC). MARCKSL1 (MARCKS-like protein 1), also known as MacMARCKS, MLP, MRP, or F52, is a 195 amino acid protein that, like MARCKS, is a major substrate for PKC. Expressed in a variety of tissues with highest levels found in testis and uterus, MARCKSL1 participates in the coordination of membrane-cytoskeletal signaling events, including secretion, migration, phagocytosis and cell adhesion. Additionally, MARCKSL1 functions as a regulator of Integrin activation and is thought to regulate Integrin-dependent signal transduction pathways, especially those involved in macrophage spreading.
Specifications
| MARCKSL1 | |
| Polyclonal | |
| Unconjugated | |
| Marcksl1 | |
| AL022768; AW215397; AW536807; brain protein F52; D4Bc1; F52; MACMARCKS; Mac-MARCKS; Macrophage myristoylated alanine-rich C kinase substrate; Macs2; Macs3; MARCKS like 1; Marcksl1; MARCKS-like 1; MARCKS-like protein 1; MARCKS-related protein; MLP; MLP1; Mrp; myristoylated alanine-rich C kinase substrate | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 17357, 65108, 81520 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P28667, P49006, Q9EPH2 | |
| Marcksl1 | |
| A synthesized peptide derived from human MARCKSL1(Accession P49006), corresponding to amino acid residues E84-C134. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction