missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARCKSL1, Mouse anti-Human, Clone: 8C8, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00065108-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: RNRKEGGGDSSASSPTEEEQEQGEIGACSDEGTAQEGKAAATPESQEPQAKGAEASAASEEEAGPQATEPSTPSGPESGPTPASAEQNESpecifications
| MARCKSL1 | |
| Monoclonal | |
| Unconjugated | |
| MARCKS-like 1 | |
| F52/MACMARCKS/MLP/MLP1/MRP | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 8C8 | |
| In 1x PBS, pH 7.4 | |
| NM_023009 | |
| MARCKSL1 | |
| MARCKSL1 (NP_075385, 107 a.a. ∼ 195 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 65108 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction