missing translation for 'onlineSavingsMsg'
Learn More

MARCKS like protein Antibody (8C8), Novus Biologicals™

Product Code. 18408389 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408389 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408389 Supplier Novus Biologicals Supplier No. H00065108M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MARCKS like protein Monoclonal antibody specifically detects MARCKS like protein in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen MARCKS like protein
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Sandwich ELISA
Classification Monoclonal
Clone 8C8
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_075385
Gene Alias F52, mac-MARCKS, MacMARCKS, Macrophage myristoylated alanine-rich C kinase substrate, MARCKS-like 1, MARCKS-like protein 1, MARCKS-related protein, MLP1, MLPMARCKS-like protein, MRPMACMARCKS
Host Species Mouse
Immunogen MARCKSL1 (NP_075385, 107 a.a. ∽ 195 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. RNRKEGGGDSSASSPTEEEQEQGEIGACSDEGTAQEGKAAATPESQEPQAKGAEASAASEEEAGPQATEPSTPSGPESGPTPASAEQNE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 65108
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.