missing translation for 'onlineSavingsMsg'
Learn More

MARCKS Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18428619 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18428619 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18428619 Supplier Novus Biologicals Supplier No. NBP305539AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MARCKS Monoclonal antibody specifically detects MARCKS in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen MARCKS
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias 80K-L, 80K-L protein, FLJ14368, MACSmyristoylated alanine-rich C-kinase substrate, MRACKS, myristoylated alanine-rich protein kinase C substrate, myristoylated alanine-rich protein kinase C substrate (MARCKS, 80K-L), phosphomyristin, PKCSLFLJ90045, PRKCSL, Protein kinase C substrate, 80 kDa protein, light chain
Host Species Mouse
Immunogen Full length human MARCKS expressed in and purified from E. coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4082
Target Species Human, Monkey
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.