missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MARCH9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103817
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MARCH9.
MARCH9 is a member of the MARCH family of membrane-bound E3 ubiquitin ligases (EC 6.3.2.19). MARCH enzymes add ubiquitin to target lysines in substrate proteins, thereby signaling their vesicular transport between membrane compartments. MARCH9 induces internalization of several membrane glycoproteins and directs them to the endosomal compartment.
Specifications
| MARCH9 | |
| Polyclonal | |
| Unconjugated | |
| MARCH9 | |
| 9-Mar; AI608492; BC019560; E3 ubiquitin-protein ligase MARCH9; MARCH9; MARCH-IX; membrane associated ring finger 9; membrane associated ring-CH-type finger 9; membrane-associated ring finger (C3HC4) 9; membrane-associated ring finger 9; membrane-associated RING finger protein 9; Membrane-associated RING-CH protein IX; RING finger protein 179; RING-type E3 ubiquitin transferase MARCH9; RNF179 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 216438, 92979 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q3TZ87, Q86YJ5 | |
| MARCH9 | |
| A synthesized peptide derived from human MARCH9(Accession Q86YJ5), corresponding to amino acid residues C46-S96. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction