missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARCH8 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | MARCH8 |
|---|---|
| Concentration | 0.13 mg/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16872614
|
Proteintech
14119-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16862614
|
Proteintech
14119-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MARCH8 (c-MIR) is a novel E3 ubiquitin ligase designated as the modulator of immune recognition (MIR) family, whose catalytic domain is a variant RING domain (RING-CH domain). MARCH8 was found as a functional and structural homolog of KSHV MIR1 and MIR2. MARCH8 targets B7-2 to lysosomal degradation and down-regulates the B7-2 surface expression through ubiquitination of its cytoplasmic tail. Furthermore, MARCH8 has been shown to down-regulate the expression of transferrin receptor and Fas, an important molecule for the induction of apoptosis. MARCH8 is the first example of an E3 ubiquitin ligase that is capable of inhibiting MHC II expression. Recent findings suggest that MARCH8 may regulate immune responses by promoting ubiquitination of MHC-II and CD86, leading to their subsequent endocytosis and lysosomal degradation.Specifications
| MARCH8 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q5T0T0, Q9DBD2 | |
| 220972, 71779 | |
| MARCH8 Fusion Protein Ag5268 | |
| Primary | |
| -20°C | |
| MARCH8 |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| c MIR, MARCH VIII, MARCH8, MIR, RING finger protein 178, RNF178 | |
| MARCH8 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title