missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARCH6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-58931
This item is not returnable.
View return policy
Description
44261 Polyclonal specifically detects 44261 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| MARCH6 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| E3 ubiquitin-protein ligase MARCH6, EC 6.3.2.-, MARCH-VIDOA10, membrane-associated ring finger (C3HC4) 6, Membrane-associated RING finger protein 6, Membrane-associated RING-CH protein VI, Protein TEB-4, RNF176KIAA0597Doa10 homolog, TEB4RING finger protein 176 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| MARCHF6 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:HTRQWLKGLVRAWTVTAGYLLDLHSYLLGDQEENENSANQQVNNNQHARNNNAIPVVGEGLHAAHQAILQQGGPVGFQPYRRPLN | |
| 100 μL | |
| Zinc Finger | |
| 10299 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction