missing translation for 'onlineSavingsMsg'
Learn More

MARCH6 Antibody (2F7), Novus Biologicals™

Product Code. 18343029 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18343029 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18343029 Supplier Novus Biologicals Supplier No. H00010299M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MARCH6 Monoclonal antibody specifically detects MARCH6 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen MARCH6
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2F7
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_005876
Gene Alias E3 ubiquitin-protein ligase MARCH6, EC 6.3.2.-, MARCH-VIDOA10, membrane-associated ring finger (C3HC4) 6, Membrane-associated RING finger protein 6, Membrane-associated RING-CH protein VI, Protein TEB-4, RNF176KIAA0597Doa10 homolog, TEB4RING finger protein 176
Host Species Mouse
Immunogen MARCH6 (NP_005876, 2 a.a. ~ 91 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. DTAEEDICRVCRSEGTPEKPLYHPCVCTGSIKFIHQECLVQWLKHSRKEYCELCKHRFAFTPIYSPDMPSRLPIQDIFAGLVTSIGTAIR
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 10299
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.