missing translation for 'onlineSavingsMsg'
Learn More

MARCH5 Antibody (3C3), Novus Biologicals™

Product Code. 18323359 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18323359 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18323359 Supplier Novus Biologicals Supplier No. H00054708M06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MARCH5 Monoclonal antibody specifically detects MARCH5 in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen MARCH5
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 3C3
Conjugate Unconjugated
Dilution ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_060294
Gene Alias E3 ubiquitin-protein ligase MARCH5, EC 6.3.2, EC 6.3.2.-, FLJ20445, MARCH-VRNF153, membrane-associated ring finger (C3HC4) 5, Membrane-associated RING finger protein 5, Membrane-associated RING-CH protein V, Mitochondrial ubiquitin ligase, RING finger protein 153MITOL
Host Species Mouse
Immunogen MARCH5 (NP_060294.1, 1 a.a. ~ 95 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MPDQALQQMLDRSCWVCFATDEDDRTAEWVRPCRCRGSTKWVHQACLQRWVDEKQRGNSTARVACPQCNAEYLIVFPKLGPVVYVLDLADRLISK
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 54708
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.