missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARCH4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24502SS
This item is not returnable.
View return policy
Description
44259 Polyclonal specifically detects 44259 in Human, Mouse, Rat, Equine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| MARCH4 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.2% Gelatin with 0.05% Sodium Azide | |
| 43163 | |
| A portion of amino acids 300-350 of human RNF174 was used as the immunogen. | |
| 0.025 mg | |
| Zinc Finger | |
| 57574 | |
| Human, Mouse, Rat, Equine, Primate | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 0.5-2 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml | |
| EC 6.3.2.-, KIAA1399E3 ubiquitin-protein ligase MARCH4, MARCH-IVMGC104908, membrane-associated ring finger (C3HC4) 4, Membrane-associated RING finger protein 4, Membrane-associated RING-CH protein IV, RNF174RING finger protein 174 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, CRA_b, CRA_c and CRA_d) and chimpanzee, and 88% homologous in monkey. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction