missing translation for 'onlineSavingsMsg'
Learn More

MARCH3 Antibody (1F6), Novus Biologicals™

Product Code. 18419409 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419409 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419409 Supplier Novus Biologicals Supplier No. H00115123M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MARCH3 Monoclonal antibody specifically detects MARCH3 in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen MARCH3
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1F6
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_848545
Gene Alias EC 6.3.2, EC 6.3.2.-, MARCH-IIIFLJ20096, membrane-associated ring finger (C3HC4) 3, Membrane-associated RING finger protein 3, Membrane-associated RING-CH protein III, MGC48332, RING finger protein 173, RNF173E3 ubiquitin-protein ligase MARCH3
Host Species Mouse
Immunogen MARCH3 (NP_848545, 1 a.a. ∽ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MTTSRCSHLPEVLPDCTSSAAPVVKTVEDCGSLVNGQPQYVMQVSAKDGQLLSTVVRTLATQSPFNDRPMCRICHEGSSQEDLLSPCECTGTLGTIHRSC
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 115123
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.