missing translation for 'onlineSavingsMsg'
Learn More

MARCH2 Antibody, Novus Biologicals™

Product Code. 18176195 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18176195 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18176195 Supplier Novus Biologicals Supplier No. NBP221582

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

44257 Polyclonal specifically detects 44257 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MARCH2
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9P0N8
Gene Alias EC 6.3.2, EC 6.3.2.-, MARCH-IIHSPC240, membrane-associated ring finger (C3HC4) 2, Membrane-associated RING finger protein 2, Membrane-associated RING-CH protein II, RING finger protein 172, RNF172E3 ubiquitin-protein ligase MARCH2
Gene Symbols 43161
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human MARCH2. The exact sequence is proprietary.
Molecular Weight of Antigen 27 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 51257
Test Specificity Human. Expected to cross react based on sequence identity: Rat (94%), Xenopus laevis (80%), Bovine (91%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.