missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MARCH1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569223
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
E3 ubiquitin-protein ligase that mediates ubiquitination of TFRC, CD86, FAS and MHC class II proteins, such as HLA-DR alpha and beta, and promotes their subsequent endocytosis and sorting to lysosomes via multivesicular bodies. By constitutively ubiquitinating MHC class II proteins in immature dendritic cells, down-regulates their cell surface localization thus sequestering them in the intracellular endosomal system.
Specifications
| MARCH1 | |
| Polyclonal | |
| Unconjugated | |
| MARCH1 | |
| 1-Mar; 2900024D24Rik; BB085186; E3 ubiquitin-protein ligase MARCH1; MARCH1; MARCHF1; MARCH-I; membrane associated ring finger 1; membrane associated ring-CH-type finger 1; membrane-associated ring finger (C3HC4) 1; membrane-associated ring finger (C3HC4) 1, E3 ubiquitin protein ligase; membrane-associated RING finger protein 1; Membrane-associated RING-CH protein I; RGD1305148; RING finger protein 171; RING-type E3 ubiquitin transferase MARCH1; RNF171 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 55016 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Surface Plasmon Resonance | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8TCQ1 | |
| MARCH1 | |
| synthetic peptide directed towards the N terminal region of human MARCH1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction