missing translation for 'onlineSavingsMsg'
Learn More

Marcasite, naturally occurring mineral, grains, ≈0.06 to 0.19 in., Thermo Scientific™

Product Code. 11379538
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379538 1 g 1g
11389538 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11379538 Supplier Thermo Scientific Alfa Aesar Supplier No. 043363.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-36-0
Molecular Formula FeS2
Molecular Weight (g/mol) 119.965
MDL Number MFCD00064690
InChI Key NFMAZVUSKIJEIH-UHFFFAOYSA-N
Synonym iron disulfide, iron sulfide, pyrite, bis sulfanylidene iron, disulfanylideneiron, iron disulfide iron pyrite, marcasite, naturally occuring mineral, grains, approximately 0.06-0.19 in, pyrite, naturally occurring mineral, grains, approximately 0.06-0.19in
PubChem CID 14788
IUPAC Name bis(sulfanylidene)iron
SMILES S=[Fe]=S

Specifications

Quantity 1 g
Solubility Information Insoluble in water.
Packaging Glass bottle
Chemical Name or Material Marcasite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.