missing translation for 'onlineSavingsMsg'
Learn More

Marbofloxacin, MedChemExpress

Product Code. 16468116
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16468116

Brand: Medchem Express HYB0126

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Marbofloxacin is a third generation fluoroquinolone and orally active antimicrobial agent, which has a broad spectrum bactericidal activity and good efficacy. Marbofloxacin can be used for the research of infections by Gram-positive and Gram-negative bacteria and Mycoplasma.

Marbofloxacin is a third generation fluoroquinolone and orally active antimicrobial agent, which has a broad spectrum bactericidal activity and good efficacy. Marbofloxacin can be used for the research of infections by Gram-positive and Gram-negative bacteria and Mycoplasma.

Specifications

Chemical Name or Material Marbofloxacin
Physical Form Solid
CAS 115550-35-1
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C17H19FN4O4
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 3.8 mg/mL (10.49 mM)
SMILES O=C(C(C1=O)=CN2N(C)COC3=C(N4CCN(C)CC4)C(F)=CC1=C32)O
Molecular Weight (g/mol) 362.36
Formula Weight 362.36
Percent Purity 99.77%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.