missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Marbofloxacin, Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Expert Technical Support
Specifications
Specifications
| Chemical Name or Material | Marbofloxacin |
| Packaging | Glass Bottle |
| CAS | 115550-35-1 |
| Color | Light Yellow |
| Melting Point | 268°C |
| Synonym | 9-Fluoro-2,3-dihydro-3-methyl-10-(4-methyl-piperazino)-7-oxo-7H-pyrido[1,2,3-ij][1,2,4]benzoxadiazine-6-carboxylic acid |
| Industry Type | Environmental Analysis |
| Molecular Weight (g/mol) | 362.36 |
| Quantity | 100 mg |
| Grade | Analytical Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction