missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Marble Chips, Technical, Granule Size 20-30mm, Fisher Chemical™
Chemical Identifiers
CAS | 471-34-1 |
---|---|
Molecular Formula | CCaO3 |
Molecular Weight (g/mol) | 100.09 |
MDL Number | MFCD00010906 |
InChI Key | VTYYLEPIZMXCLO-UHFFFAOYSA-L |
IUPAC Name | calcium carbonate |
SMILES | [Ca++].[O-]C([O-])=O |
Description
Chemical Identifiers
471-34-1 | |
100.09 | |
VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
[Ca++].[O-]C([O-])=O |
CCaO3 | |
MFCD00010906 | |
calcium carbonate |
Specifications
825°C | |
White | |
Solid | |
MFCD00010906 | |
[Ca++].[O-]C([O-])=O | |
100.09 | |
Plastic powder jar |
471-34-1 | |
8 to 9 | |
CCaO3 | |
VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
calcium carbonate | |
100.09g/mol |