missing translation for 'onlineSavingsMsg'
Learn More

Maraviroc, MedChemExpress

Product Code. 16481647
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481647 50 mg 50mg
16458187 5 mg 5mg
16416066 100 mg 100mg
16405478 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16481647 Supplier Medchem Express Supplier No. HY13004

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Maraviroc (UK-427857) is a selective CCR5 antagonist with activity against human HIV.

Maraviroc (UK-427857) is a selective CCR5 antagonist with activity against human HIV.

Specifications

Chemical Name or Material Maraviroc
CAS 376348-65-1
Color White
Purity Grade Notes Research
Molecular Formula C29H41F2N5O
Quantity 50 mg
Synonym UK-427857
Solubility Information DMSO : 50 mg/mL (97.34 mM; Need ultrasonic) ∣Ethanol : 6.5 mg/mL (12.65 mM; Need ultrasonic)
SMILES O=C(C1CCC(F)(F)CC1)N[C@H](C2=CC=CC=C2)CCN3[C@H]4C[C@@H](N5C(C)=NN=C5C(C)C)C[C@@H]3CC4
Molecular Weight (g/mol) 513.67
Formula Weight 513.67
Percent Purity 98.18%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.