Learn More
Invitrogen™ Marapsin/PRSS27 Human ELISA Kit
Sandwich ELISA Kit
£544.00 - £4280.00
Specifications
Accession Number | Q9BQR3 |
---|---|
Assay Sensitivity | 35 pg/mL |
Assay | Solid-phase sandwich ELISA |
Conjugate | Biotin |
Product Type | ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15465113
|
Invitrogen™
EHPRSS27 |
96 Tests |
£544.00
1 set |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
17804220
|
Invitrogen™
EHPRSS27X10 |
10 x 96 Tests |
£4280.00
Pack of 10 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
17165549
|
Invitrogen™
EHPRSS27X5 |
5 x 96 Tests |
£2320.00
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
The Human Marapsin/PRSS27 ELISA quantitates Hu Marapsin in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Marapsin. Principle of the method The Human Marapsin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Pancreasin is a pancreatic tryptic serine peptidase that cleaves peptides after an arginine residue., Sep 2003].Specifications
Q9BQR3 | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
Human | |
Colorimetric Microplate Reader | |
PRSS27 | |
<10% | |
RUO | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
35 pg/mL | |
Biotin | |
Cell Structure & Organelle Proteins | |
This ELISA kit shows no cross-reactivity with the following cytokines tested: human Angiogenin, BDNF, BLC, CNTF, ENA- 78, FGF-4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL- 11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, FGF-4, FGF-6, FGF- 7, G-CSF, GDNF, GM-CSF, IFN-gamma, IGFBP-2, IGFBP-3, IGFBP-4, Leptin (OB), MCP-1, MCP-2, MCP-3, MDC, MIF, MIG, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, SDF-1 alpha, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF. | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
83886 | |
<12% | |
HRP | |
Cell Cycle & Proliferation | |
UNQ1884/PRO4327, CAPH2, MPN | |
1 hr. 20 min. |