missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Marapsin/Pancreasin Rabbit anti-Human, Clone: 109, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90057-100UL
This item is not returnable.
View return policy
Description
Marapsin/Pancreasin Monoclonal specifically detects Marapsin/Pancreasin in Human samples. It is validated for ELISA.
Specifications
| Marapsin/Pancreasin | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 83886 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| 109 | |
| ELISA 1:5000-1:10000 | |
| CAPH2, channel-activating protease 2, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, EC 3.4.21.43, marapsin, MPNMarapsin, Pancreasin, protease, serine 27, serine protease 27 | |
| Recombinant Human Marapsin/Pancreasin Protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction