missing translation for 'onlineSavingsMsg'
Learn More

Marapsin/Pancreasin Antibody (MM0463-3H15), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18430598 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430598 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430598 Supplier Novus Biologicals Supplier No. NBP211786MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Marapsin/Pancreasin Monoclonal antibody specifically detects Marapsin/Pancreasin in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Marapsin/Pancreasin
Applications Western Blot
Classification Monoclonal
Clone MM0463-3H15
Conjugate mFluor Violet 500 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias CAPH2, channel-activating protease 2, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, EC 3.4.21.43, marapsin, MPNMarapsin, Pancreasin, protease, serine 27, serine protease 27
Host Species Mouse
Immunogen Human recombinant Marapsin/Pancreasin
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83886
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.