Learn More
Marapsin/Pancreasin Antibody (MM0463-3H15), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-11786JF549
Product Code. 18454177
  • £486.00 / 0.10mL
Description
Marapsin/Pancreasin Monoclonal antibody specifically detects Marapsin/Pancreasin in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Marapsin/Pancreasin
Monoclonal
Janelia Fluor 549
50mM Sodium Borate
Mouse
Protein G purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
MM0463-3H15
Western Blot
CAPH2, channel-activating protease 2, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, EC 3.4.21.43, marapsin, MPNMarapsin, Pancreasin, protease, serine 27, serine protease 27
Human recombinant Marapsin/Pancreasin
0.1 mL
Cancer, Inflammation, Innate Immunity, Necroptosis, Neurodegeneration, Signal Transduction
83886
Store at 4C in the dark.
IgG1