missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Maprotiline Hydrochloride, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Maprotiline Hydrochloride |
| CAS | 10347-81-6 |
| Analyte or Component Names | Maprotiline Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H23 N . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | 9,10-Ethanoanthracene-9(10H)-propanamine, N-methyl-, hydrochloride (1:1), 9,10-Ethanoanthracene-9(10H)-propanamine, N-methyl-, hydrochloride (9CI), 9,10-Ethanoanthracene-9(10H)-propylamine, N-methyl-, hydrochloride (7CI,8CI), 9,10-Dihydro-9-(3-methylaminopropyl)-9,10-ethanoanthracene hydrochloride, 9,10-Ethanoanthracene-9-propylamine, 9,10-dihydro-N-methyl-, hydrochloride, Ba 34276, CIBA 34,276Ba, CIBA 34276 hydrochloride, Deprilept, Ladiomil, Ludiomil, Maprotiline hydrochloride, Psymion, 9-[gamma-(Methylamino)propyl]-9,10-dihydro-9,10-ethanoanthracene hydrochloride |
| InChI Formula | InChI=1S/C20H23N.ClH/c1-21-14-6-12-20-13-11-15(16-7-2-4-9-18(16)20)17-8-3-5-10-19(17)20;/h2-5,7-10,15,21H,6,11-14H2,1H3;1H |
| Solution Type | Neat |
| SMILES | Cl.CNCCCC12CCC(c3ccccc13)c4ccccc24 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction