missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAPRE3 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA519254
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine and canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
EB3 is a 282 amino acid homologue of EB1, a protein known to bind APC (adenomatous polyposis coli). It has been shown to bind to tubulin in vitro and microtubules in vivo. Northern blotting shows expressions to be mainly in brain, and to a lesser degree in muscle. EB3 interacts with APCL, a homologue of the tumor suppressor APC, and this interaction may be involved in the stabilization or extension of microtubules.
Specifications
| MAPRE3 | |
| Polyclonal | |
| Unconjugated | |
| MAPRE3 | |
| AI790651; APC binding protein; EB1 protein family member 3; EB2; EB3; EBF3; EBF3-S; End-binding protein 3; Mapre3; microtubule associated protein RP/EB family member 3; microtubule-associated protein RP/EB family member 3; microtubule-associated protein, RP/EB family, member 3; RP3 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 100732, 22924 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q6PER3, Q9UPY8 | |
| MAPRE3 | |
| Synthetic peptide sequence (SGRLSNVAPPCILR) corresponding to the internal amino acids of MAPRE3 (aa 172-185). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction