Learn More
MAPRE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10364-1-AP-20UL
 View more versions of this product
Product Code. 16866703
  • £135.00 / 20µL
Description
The protein encoded by this gene shares significant homology to the adenomatous polyposis coli (APC) protein-binding EB1 gene family. The function of this protein is unknown; however, its homology suggests involvement in tumorigenesis of colorectal cancers and proliferative control of normal cells. This gene may belong to the intermediate/early gene family, involved in the signal transduction cascade downstream of the TCR. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
MAPRE2
Polyclonal
Unconjugated
MAPRE2
APC binding protein EB2, EB1, EB2, End binding protein 2, MAPRE2, RP1
Rabbit
Antigen Affinity Chromatography
RUO
10982, 212307, 679221
-20°C
Liquid
Immunocytochemistry, Western Blot, Immunofluorescence
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q15555, Q3B8Q0, Q8R001
Mapre2
MAPRE2 Fusion Protein Ag0416
20 μL
Primary
Human, Rat, Mouse