missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPKAPK3, Mouse anti-Human, Clone: 1C3, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00007867-M04.100ug
This item is not returnable.
View return policy
Description
Sequence: LECFHRRTGQKCALKLLYDSPKARQEVDHHWQASGGPHIVCILDVYENMHHGKRCLLIIMECMEGGELFSRIQERGDQAFTEREAAEIMRDIGTASpecifications
| MAPKAPK3 | |
| Monoclonal | |
| Unconjugated | |
| mitogen-activated protein kinase-activated protein kinase 3 | |
| 3PK/MAPKAP3 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1C3 | |
| In 1x PBS, pH 7.4 | |
| NM_004635 | |
| MAPKAPK3 | |
| MAPKAPK3 (NP_004626, 59 a.a. ∼ 153 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 7867 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction