Learn More
MAPKAPK2 (phospho S272) Rabbit anti-Human, Mouse, Rabbit, Polyclonal , Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human MAPKAPK2.
Brand: Abnova PAB29630.100uL
Description
This gene encodes a member of the Ser/Thr protein kinase family. This kinase is regulated through direct phosphorylation by p38 MAP kinase. In conjunction with p38 MAP kinase, this kinase is known to be involved in many cellular processes including stress and inflammatory responses, nuclear export, gene expression regulation and cell proliferation. Heat shock protein HSP27 was shown to be one of the substrates of this kinase in vivo. Two transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq]
Specifications
| MAPKAPK2 (phospho S272) | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of human MAPKAPK2. | |
| In PBS (without Mg2+ and Ca2+), 150 mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| MAPKAPK2 | |
| A synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S272 of human MAPKAPK2. | |
| 100 μL | |
| Primary | |
| MAPKAPK2 (phospho S272) polyclonal antibody detects endogenous level of MAPKAPK2 only when phosphorylated at serine 272. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| MK2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 9261 | |
| Human, Mouse, Rabbit | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.