missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MAPKAPK2 Monoclonal antibody specifically detects MAPKAPK2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | MAPKAPK2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 7H4.2 |
| Conjugate | DyLight 594 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 2.7.11, EC 2.7.11.1, MAP kinase-activated protein kinase 2, MAPK-activated protein kinase 2, MAPKAP kinase 2, MAPKAPK-2, mitogen-activated protein kinase-activated protein kinase 2, MK2 |
| Host Species | Mouse |
| Immunogen | KLH-conjugated synthetic peptide of the C-terminus of the long form of human MAPKAP-K2. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?