missing translation for 'onlineSavingsMsg'
Learn More

MAPKAPK2 Antibody (7H4.2), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18403248 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.2 mL
Unit Size:
0.2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18403248 0.2 mL 0.2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18403248 Supplier Novus Biologicals Supplier No. NBP229767AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MAPKAPK2 Monoclonal antibody specifically detects MAPKAPK2 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAPKAPK2
Applications Western Blot
Classification Monoclonal
Clone 7H4.2
Conjugate Alexa Fluor 594
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.11, EC 2.7.11.1, MAP kinase-activated protein kinase 2, MAPK-activated protein kinase 2, MAPKAP kinase 2, MAPKAPK-2, mitogen-activated protein kinase-activated protein kinase 2, MK2
Host Species Mouse
Immunogen KLH-conjugated synthetic peptide of the C-terminus of the long form of human MAPKAP-K2.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, MAP Kinase Signaling, Phospho Specific, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9261
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.