missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX MAPKAP Kinase 3 Protein, Unactive

Product Code. 17380384
Encompass_Preferred
Click to view available options
Quantity:
250 μg
50 μg
Unit Size:
250µg
50µg
This item is not returnable. View return policy

Product Code. 17380384

Brand: Eurofins DiscoverX 14586

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Unactive, N-terminal, GST-tagged, recombinant, human MAPKAP Kinase 3, amino acids 2-end, for use in Kinase Assays.

MAPKAP Kinase 3 is a stress-activated serine/threonine-protein kinase which involved in cytokines production, endocytosis, cell migration, chromatin remodeling and transcriptional regulation. This kinase functions as a mitogen-activated protein kinase (MAP kinase)- activated protein kinase. MAP kinases are also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals. This kinase was shown to be activated by growth inducers and stress stimulation of cells. In vitro studies demonstrated that ERK, p38 MAP kinase and Jun N-terminal kinase were all able to phosphorylate and activate this kinase, which suggested the role of this kinase as an integrative element of signaling in both mitogen and stress responses. This kinase was reported to interact with, phosphorylate and repress the activity of E47, which is a basic helix-loop-helix transcription factor known to be involved in the regulation of tissue-specific gene expression and cell differentiation.

GenBank NM_004635

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Molecular Weight (g/mol) 69.8 kDa
Quantity 50 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal GST
Expression System Recombinant enzyme expressed in E. coli cells
Protein Form Truncated
Purity or Quality Grade ≥68%
Protein MAPKAP Kinase 3, unactive
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.