missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPKAP Kinase 3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-37960-100ul
This item is not returnable.
View return policy
Description
MAPKAP Kinase 3 Polyclonal antibody specifically detects MAPKAP Kinase 3 in Human,Mouse samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| MAPKAP Kinase 3 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| 3PKEC 2.7.11.1, Chromosome 3p kinase, EC 2.7.11, MAP kinase-activated protein kinase 3, MAPK-activated protein kinase 3, MAPKAP kinase 3, MAPKAP33pK, MAPKAPK-3, mitogen-activated protein kinase-activated protein kinase 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human MAPKAP Kinase 3 (NP_004626.1).,, Sequence:, MDGETAEEQGGPVPPPVAPGGPGLGGAPGGRREPKKYAVTDDYQLSKQVLGLGVNGKVLECFHRRTGQKCALKLLYDSPKARQEVDHHWQASGGPHIVCILDVYENMHHGKRCLLIIMECMEGGELFSRIQERGDQAFTEREAAEIMRDIGTAIQFLHSHNIAHRDVKPENLLYTSKEKDAVLKLTDFGFAKETTQNALQTPCYTPYYVAPEVLGPEKYDKSCDMWSLGVIMYILLCGFP | |
| 100 μL | |
| Protein Kinase | |
| 7867 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction