missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAPK4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103300
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total p63 MAPK.
Mitogen-activated protein kinase 4 is a member of the mitogen-activated protein kinase family. Tyrosine kinase growth factor receptors activate mitogen-activated protein kinases which then translocate into the nucleus where it phosphorylates nuclear targets.
Specifications
| MAPK4 | |
| Polyclonal | |
| Unconjugated | |
| MAPK4 | |
| A330097D03Rik; Erk3; Erk3-related; ERK4; ERK-4; Extracellular signal-regulated kinase 4; MAP kinase 4; MAP kinase isoform p63; MAPK 4; MAPK4; mitogen activated protein kinase 4; mitogen-activated protein kinase 4; MNK2; p63Mapk; p63-MAPK; Prkm4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 225724, 5596 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P31152, Q6P5G0 | |
| MAPK4 | |
| A synthesized peptide derived from human MAPK4(Accession P31152), corresponding to amino acid residues Q344-Q394. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction