missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAPK15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528587
This item is not returnable.
View return policy
Description
Recommended positive controls: HeLaS3. Predicted reactivity: Human (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The ERKs are a subfamily of the MAPKs that have been implicated in cell growth and differentiation. Extracellular signal-regulated kinase 8 (Erk8) is a large MAP kinase whose activity is controlled by serum and the c-Src non-receptor tyrosine kinase. ERK8 down-regulates transactivation of the glucocorticoid receptor through Hic-5 and can negatively regulate transcriptional co-activation of androgen receptor and GRalpha by Hic-5 in a kinase-independent manner, suggesting a broader role for ERK8 in the regulation of nuclear receptors beyond estrogen receptor alpha. Erk8 is a novel effector of RET/PTC3 and, therefore, RET biological functions.
Specifications
| MAPK15 | |
| Polyclonal | |
| Unconjugated | |
| MAPK15 | |
| BC048082; EC 2.7.11.24; ERK7; ERK-7; ERK8; ERK-8; extracellular regulated kinase 8 delta; extracellular signal regulated kinase 8; extracellular signal-regulated kinase 7; Extracellular signal-regulated kinase 8; MAP kinase 15; MAPK 15; Mapk15; Mitogen-activated protein kinase 15 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 225689 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q8TD08 | |
| MAPK15 | |
| Recombinant protein encompassing a sequence within the center region of human MAPK15. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction