missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPK13 Mouse anti-Human, Mouse, Clone: 1B12A6, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | MAPK13 |
|---|---|
| Clone | 1B12A6 |
| Concentration | 0.33 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16827295
|
Proteintech
60252-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16817295
|
Proteintech
60252-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is a member of the MAP kinase family. MAP kinases act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. This kinase is closely related to p38 MAP kinase, both of which can be activated by proinflammatory cytokines and cellular stress. MAP kinase kinases 3, and 6 can phosphorylate and activate this kinase. Transcription factor ATF2, and microtubule dynamics regulator stathmin have been shown to be the substrates of this kinase.Specifications
| MAPK13 | |
| 0.33 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| MAP kinase 13, MAP kinase p38 delta, MAPK 13, MAPK13, p38delta, PRKM13, SAPK4 | |
| MAPK13 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1B12A6 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| O15264, Q9Z1B7 | |
| 26415, 5603 | |
| MAPK13 Fusion Protein Ag21240 | |
| Primary | |
| -20°C | |
| MAPK13 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title