missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAP7D1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544693
This item is not returnable.
View return policy
Description
Peptide sequence: RRRLEEQRLK AEQRRAALEE RQRQKLEKNK ERYEAAIQRS VKKTWAEIRQ Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
MAP7D1 (MAP7 Domain Containing 1) is a Protein Coding gene.
Specifications
| MAP7D1 | |
| Polyclonal | |
| Unconjugated | |
| Map7d1 | |
| arginine/proline rich coiled-coil 1; arginine/proline-rich coiled-coil domain-containing protein 1; AV028413; BC019977; FLJ10350; FLJ39022; KIAA1187; MAP7 domain containing 1; MAP7 domain-containing protein 1; MAP7D1; microtubule-associated protein 7 domain containing 1; mKIAA1187; Mtap7d1; Parcc1; PP2464; proline arginine rich coiled coil 1; proline/arginine-rich coiled-coil domain-containing protein 1; Rprc1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 245877, 55700 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| A2AJI0, Q3KQU3 | |
| Map7d1 | |
| Synthetic peptide directed towards the N-terminal of human MAP7D1 (aa 179-228). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction