Learn More
MAP7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13446-1-AP-150UL
 View more versions of this product
Product Code. 16891664
  • £349.00 / 150µL
Description
The product of this gene is a microtubule-associated protein that is predomitly expressed in cells of epithelial origin. Microtubule-associated proteins are thought to be involved in microtubule dynamics, which is essential for cell polarization and differentiation. This protein has been shown to be able to stabilize microtubules, and may serve to modulate microtubule functions. Studies of the related mouse protein also suggested an essential role in microtubule function required for spermatogenesis. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
MAP7
Polyclonal
Unconjugated
MAP7
E MAP 115, EMAP115, Ensconsin, MAP 7, MAP7
Rabbit
Antigen Affinity Chromatography
RUO
17761, 293016, 9053
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.4 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O88735, Q14244
Map7
MAP7 Fusion Protein Ag4276
150 μL
Primary
Human, Mouse, Rat